missing translation for 'onlineSavingsMsg'
Learn More

Honokiol, TRC

Product Code. 30395663
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395663 50 mg
30390316 25 mg
30407456 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395663 Supplier Toronto Research Chemicals Supplier No. TRCH66956050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35354-74-6
Molecular Formula C18 H18 O2
Molecular Weight (g/mol) 266.33
Synonym 3',5-Di-2-propen-1-yl-[1,1'-biphenyl]-2,4'-diol, [1,1'-Biphenyl]-2,4'-diol, 3',5-di-2-propenyl- (9CI), 3',5-Diallyl-2,4'-biphenyldiol, Honokiol, NSC 293100
IUPAC Name 2-(4-hydroxy-3-prop-2-enylphenyl)-4-prop-2-enylphenol
SMILES Oc1ccc(cc1CC=C)c2cc(CC=C)ccc2O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H18O2/c1-3-5-13-7-9-18(20)16(11-13)14-8-10-17(19)15(12-14)6-4-2/h3-4,7-12,19-20H,1-2,5-6H2
Formula Weight 266.1307
Chemical Name or Material Honokiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.