missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-dimethylbenzoic Acid Methyl Ester, TRC

Product Code. 30396212
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396212 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396212 Supplier Toronto Research Chemicals Supplier No. TRCH8253951G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxy-3,5-dimethylbenzoic Acid Methyl Ester
CAS 34137-14-9
Molecular Formula C10 H12 O3
Quantity 1 g
Synonym Benzoic acid, 4-hydroxy-3,5-dimethyl-, methyl ester, 4-Hydroxy-3,5-dimethylbenzoic acid methyl ester, Methyl 3,5-dimethyl-4-hydroxybenzoate, Methyl 4-hydroxy-3,5-dimethylbenzoate
InChI Formula InChI=1S/C10H12O3/c1-6-4-8(10(12)13-3)5-7(2)9(6)11/h4-5,11H,1-3H3
SMILES COC(=O)c1cc(C)c(O)c(C)c1
IUPAC Name methyl 4-hydroxy-3,5-dimethylbenzoate
Molecular Weight (g/mol) 180.2
Formula Weight 180.0786
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.