missing translation for 'onlineSavingsMsg'
Learn More

Isomaltotetraose, TRC

Product Code. 30399476
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399476 10 mg
30395896 25 mg
30396357 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399476 Supplier Toronto Research Chemicals Supplier No. TRCI82129010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35997-20-7
Molecular Formula C24 H42 O21
Molecular Weight (g/mol) 666.58
Synonym O-α-D-Glucopyranosyl-(1→6)-O-α-D-glucopyranosyl-(1→6)-O-α-D-glucopyranosyl-(1→6)-D-glucose
IUPAC Name (3R,4S,5S,6R)-6-[[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxan-2-yl]oxymethyl]oxan-2-yl]oxymethyl]oxane-2,3,4,5-tetrol
SMILES OC[C@H]1O[C@H](OC[C@H]2O[C@H](OC[C@H]3O[C@H](OC[C@H]4OC(O)[C@H](O)[C@@H](O)[C@@H]4O)[C@H](O)[C@@H](O)[C@@H]3O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C24H42O21/c25-1-5-9(26)14(31)18(35)22(43-5)40-3-7-11(28)16(33)20(37)24(45-7)41-4-8-12(29)15(32)19(36)23(44-8)39-2-6-10(27)13(30)17(34)21(38)42-6/h5-38H,1-4H2/t5-,6-,7-,8-,9-,10-,11-,12-,13+,14+,15+,16+,17-,18-,19-,20-,21?,22+,23+,24+/m1/s1
Formula Weight 666.2219
Chemical Name or Material Isomaltotetraose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.