missing translation for 'onlineSavingsMsg'
Learn More

Fenquinotrione, TRC

Product Code. 30400160
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400160 5 mg
30404341 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400160 Supplier Toronto Research Chemicals Supplier No. TRCF0236205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1342891-70-6
Molecular Formula C22 H17 Cl N2 O5
Molecular Weight (g/mol) 424.8338
Synonym 1,3-Cyclohexanedione, 2-[[8-chloro-3,4-dihydro-4-(4-methoxyphenyl)-3-oxo-2-quinoxalinyl]carbonyl]-, Fenquinotrione
IUPAC Name 2-[8-chloro-4-(4-methoxyphenyl)-3-oxoquinoxaline-2-carbonyl]cyclohexane-1,3-dione
SMILES COc1ccc(cc1)N2C(=O)C(=Nc3c(Cl)cccc23)C(=O)C4C(=O)CCCC4=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C22H17ClN2O5/c1-30-13-10-8-12(9-11-13)25-15-5-2-4-14(23)19(15)24-20(22(25)29)21(28)18-16(26)6-3-7-17(18)27/h2,4-5,8-11,18H,3,6-7H2,1H3
Formula Weight 424.0826
Chemical Name or Material Fenquinotrione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.