missing translation for 'onlineSavingsMsg'
Learn More

Glycerophospho-N-Arachidonoyl Ethanolamine Sodium Salt, TRC

Product Code. 30401412
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401412 25 mg -
30404670 2.5 mg 2.5mg
30404784 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401412 Supplier Toronto Research Chemicals Supplier No. TRCG59865025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C25 H43 N Na O7 P
Molecular Weight (g/mol) 523.575
Synonym (all-Z)-Phosphoric Acid Mono(2,3-dihydroxypropyl) Mono[2-[(1-oxo-5,8,11,14-eicosatetraenyl)amino]ethyl] Ester Sodium Salt, Phosphoric Acid Mono(2,3-dihydroxypropyl) Sodium Salt, vMono[2-[[(5Z,8Z,11Z,14Z)-1-oxo-5,8,11,14-eicosatetraenyl]amino]ethyl] Ester Sodium Salt
IUPAC Name [2,3-dihydroxypropoxy-[2-[[(5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoyl]amino]ethoxy]phosphoryl]oxysodium
SMILES CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)NCCOP(=O)(O[Na])OCC(O)CO

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C25H44NO7P.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-25(29)26-20-21-32-34(30,31)33-23-24(28)22-27;/h6-7,9-10,12-13,15-16,24,27-28H,2-5,8,11,14,17-23H2,1H3,(H,26,29)(H,30,31);/q;+1/p-1/b7-6-,10-9-,13-12-,16-15-;
Formula Weight 523.267
Chemical Name or Material Glycerophospho-N-Arachidonoyl Ethanolamine Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.