missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methoxycarbonylphenylboronic Acid, Pinacol Ester, TRC

Product Code. 30401676
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401676 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30401676 Supplier Toronto Research Chemicals Supplier No. TRCF5955731G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Fluoro-4-methoxycarbonylphenylboronic Acid, Pinacol Ester
CAS 603122-52-7
Molecular Formula C19 H38 F3 N O2 S
Quantity 1 g
Synonym 2-Fluoro-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzoic Acid Methyl Ester, Methyl 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
InChI Formula InChI=1S/C19H38F3NO2S/c1-15(13-17(3,4)5)9-11-23(26(24,25)19(20,21)22)12-10-16(2)14-18(6,7)8/h15-16H,9-14H2,1-8H3
SMILES CC(CCN(CCC(C)CC(C)(C)C)S(=O)(=O)C(F)(F)F)CC(C)(C)C
IUPAC Name 1,1,1-trifluoro-N,N-bis(3,5,5-trimethylhexyl)methanesulfonamide
Molecular Weight (g/mol) 401.571
Formula Weight 401.258
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.