missing translation for 'onlineSavingsMsg'
Learn More

Heptadecanedioic Acid, TRC

Product Code. 30401919
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401919 100 mg -
30398657 50 mg 50mg
30401920 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401919 Supplier Toronto Research Chemicals Supplier No. TRCH998558100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2424-90-0
Molecular Formula C17H32O4
Molecular Weight (g/mol) 300.43
Synonym 1,15-Pentadecanedicarboxylic acid, 1,17-Heptadecanedioic acid, Pentadecanedicarboxylic acid, ω-Carboxypalmitic acid
IUPAC Name heptadecanedioic acid
SMILES OC(=O)CCCCCCCCCCCCCCCC(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C17H32O4/c18-16(19)14-12-10-8-6-4-2-1-3-5-7-9-11-13-15-17(20)21/h1-15H2,(H,18,19)(H,20,21)
Formula Weight 300.2301
Chemical Name or Material Heptadecanedioic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.