missing translation for 'onlineSavingsMsg'
Learn More

3Alpha-Hydroxy Pravastatin Lactone, TRC

Product Code. 30402343
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402343 1 mg 1mg
30394650 10 mg -
30407174 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402343 Supplier Toronto Research Chemicals Supplier No. TRCH9523001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85798-96-5
Molecular Formula C23 H34 O6
Molecular Weight (g/mol) 406.51
Synonym Butanoic acid, 2-methyl-, (1S,6S,7R,8S,8aR)-1,2,6,7,8,8a-hexahydro-6-hydroxy-7-methyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, (2S)-, Butanoic acid, 2-methyl-, (1S,6S,7R,8S)-1,2,6,7,8,8a-hexahydro-6-hydroxy-7-methyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, (2S)- (9CI), SQ 31906 lactone, (2S)-2-Methylbutanoic acid (1S,6S,7R,8S,8aR)-1,2,6,7,8,8a-hexahydro-6-hydroxy-7-methyl-8-[2-[(2R,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, 3α-Hydroxy pravastatin lactone, 3α-Hydroxypravastatin lactone, 6α-Hydroxy-iso-compactin
IUPAC Name [(1S,6S,7R,8S,8aR)-6-hydroxy-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-7-methyl-1,2,6,7,8,8a-hexahydronaphthalen-1-yl] (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@H]1CC=CC2=C[C@@H](O)[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C23H34O6/c1-4-13(2)23(27)29-20-7-5-6-15-10-19(25)14(3)18(22(15)20)9-8-17-11-16(24)12-21(26)28-17/h5-6,10,13-14,16-20,22,24-25H,4,7-9,11-12H2,1-3H3/t13-,14+,16+,17+,18-,19+,20-,22-/m0/s1
Formula Weight 406.2355
Chemical Name or Material 3a-Hydroxy Pravastatin Lactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.