missing translation for 'onlineSavingsMsg'
Learn More

Haloxyfop-R-methyl-d3, TRC

Product Code. 30402530
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402530 25 mg 25mg
30399980 10 mg 10mg
30390483 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30402530 Supplier Toronto Research Chemicals Supplier No. TRCH10402225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

IUPAC Name trideuteriomethyl (2R)-2-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxyphenoxy]propanoate
SMILES [2H]C([2H])([2H])OC(=O)[C@@H](C)Oc1ccc(Oc2ncc(cc2Cl)C(F)(F)F)cc1

Specifications

Molecular Formula C16 2H3 H10 Cl F3 N O4
Quantity 25 mg
Synonym Haloxyfop-R-methyl D3 (Methoxy D3), (R)-Haloxyfop Methyl D3 (Methoxy D3)
InChI Formula InChI=1S/C16H13ClF3NO4/c1-9(15(22)23-2)24-11-3-5-12(6-4-11)25-14-13(17)7-10(8-21-14)16(18,19)20/h3-9H,1-2H3/t9-/m1/s1/i2D3
Molecular Weight (g/mol) 378.7454
Formula Weight 378.0674
Chemical Name or Material Haloxyfop-R-methyl D3 (Methoxy D3)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.