missing translation for 'onlineSavingsMsg'
Learn More

rac Fenfluramine Di-O-benzoyl-L-tartaric Acid, TRC

Product Code. 30403943
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403943 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30403943 Supplier Toronto Research Chemicals Supplier No. TRCF24760510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac Fenfluramine Di-O-benzoyl-L-tartaric Acid
CAS 895166-16-2
Molecular Formula C30H30F3NO8
Quantity 10 g
Synonym N-Ethyl-alpha-methyl-3-(trifluoromethyl)benzeneethanamine Di-O-benzoyl-L-tartaric Acid, N-Ethyl-alpha-methyl-m-(trifluoromethyl)phenethylamine Di-O-benzoyl-L-tartaric Acid, Acino Di-O-benzoyl-L-tartaric Acid, Adipomin Di-O-benzoyl-L-tartaric Acid, Obedrex Di-O-benzoyl-L-tartaric Acid, Pesos Di-O-benzoyl-L-tartaric Acid, Ponderal Di-O-benzoyl-L-tartaric Acid, Pondimin Di-O-benzoyl-L-tartaric Acid, Rotondin Di-O-benzoyl-L-tartaric Acid
InChI Formula InChI=1S/C18H14O8.C12H16F3N/c19-15(20)13(25-17(23)11-7-3-1-4-8-11)14(16(21)22)26-18(24)12-9-5-2-6-10-12;1-3-16-9(2)7-10-5-4-6-11(8-10)12(13,14)15/h1-10,13-14H,(H,19,20)(H,21,22);4-6,8-9,16H,3,7H2,1-2H3/t13-,14-;/m1./s1
SMILES CCNC(C)Cc1cccc(c1)C(F)(F)F.OC(=O)[C@H](OC(=O)c2ccccc2)[C@@H](OC(=O)c3ccccc3)C(=O)O
IUPAC Name (2R,3R)-2,3-dibenzoyloxybutanedioic acid;N-ethyl-1-[3-(trifluoromethyl)phenyl]propan-2-amine
Molecular Weight (g/mol) 589.56
Formula Weight 589.1924
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.