missing translation for 'onlineSavingsMsg'
Learn More

H-HIS-PHE-OH, TRC

Product Code. 30404704
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404704 100 mg -
30395893 10 mg -
30395111 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30404704 Supplier Toronto Research Chemicals Supplier No. TRCH388080100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16874-81-0
Molecular Formula C15H18N4O3
Molecular Weight (g/mol) 302.33
IUPAC Name (2S)-2-[[(2S)-2-amino-3-(1H-imidazol-5-yl)propanoyl]amino]-3-phenylpropanoic acid
SMILES N[C@@H](Cc1cnc[nH]1)C(=O)N[C@@H](Cc2ccccc2)C(=O)O

Specifications

InChI Formula InChI=1S/C15H18N4O3/c16-12(7-11-8-17-9-18-11)14(20)19-13(15(21)22)6-10-4-2-1-3-5-10/h1-5,8-9,12-13H,6-7,16H2,(H,17,18)(H,19,20)(H,21,22)/t12-,13-/m0/s1
Quantity 100 mg
Formula Weight 302.1379
Shipping Condition Room Temperature
Chemical Name or Material H-HIS-PHE-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.