missing translation for 'onlineSavingsMsg'
Learn More

Fenamidone Metabolite, TRC

Product Code. 30406158
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406158 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406158 Supplier Toronto Research Chemicals Supplier No. TRCF24540350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenamidone Metabolite RPA 405862
CAS 153969-11-0
Molecular Formula C16 H15 N3 O2
Quantity 50 mg
Synonym Fenamidone Metabolite, Fenamidone Metabolite RPA 405862, 5-Methyl-5-phenyl-3-(phenylamino)-2,4-imidazolidinedione (ACI), 2,4-Imidazolidinedione, 5-methyl-5-phenyl-3-(phenylamino)- (9CI, ACI)
InChI Formula InChI=1S/C16H15N3O2/c1-16(12-8-4-2-5-9-12)14(20)19(15(21)17-16)18-13-10-6-3-7-11-13/h2-11,18H,1H3,(H,17,21)
SMILES CC1(NC(=O)N(Nc2ccccc2)C1=O)c3ccccc3
IUPAC Name 3-anilino-5-methyl-5-phenylimidazolidine-2,4-dione
Molecular Weight (g/mol) 281.309
Formula Weight 281.116
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.