missing translation for 'onlineSavingsMsg'
Learn More

Phenol-2,4,6-d3, TRC

Product Code. 30409111
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409111 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30409111 Supplier Toronto Research Chemicals Supplier No. TRCP318004100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenol 2,4,6-d3
CAS 7329-50-2
Molecular Formula C6 2H3 H3 O
Quantity 100 mg
Synonym Phen-2,4,6-d3-ol, Phenol-2,4,6-d3, Phenol 2,4,6-D3, 2,4,6-Trideuteriophenol
InChI Formula InChI=1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H/i1D,4D,5D
SMILES [2H]c1cc([2H])c(O)c([2H])c1
IUPAC Name 2,4,6-trideuteriophenol
Molecular Weight (g/mol) 97.13
Formula Weight 97.0607
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.