missing translation for 'onlineSavingsMsg'
Learn More

Raubasine, TRC

Product Code. 30409658
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409658 500 mg -
30427496 250 mg 250mg
30427347 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30409658 Supplier Toronto Research Chemicals Supplier No. TRCR127000500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 483-04-5
Molecular Formula C21 H24 N2 O3
Molecular Weight (g/mol) 352.43
Synonym Oxayohimban-16-carboxylic acid, 16,17-didehydro-19-methyl-, methyl ester, (19α)-, Ajmalicine (7CI,8CI), Raubasine (6CI), 4H-Indolo[2,3-a]pyrano[3,4-g]quinolizine, oxayohimban-16-carboxylic acid deriv., (-)-Ajmalicine, 4H-Indolo[2,3-a]pyrano[3,4-g]quinolizine-1-carboxylic acid, 4a,5,7,8,13,13b,14,14a-octahydro-4-methyl-, methyl ester, (4S,4aR,13bS,14aS)-, Ajmalicin, Circolene, Hydrosarpan, Isoarteril, Lamuran, NSC 72133, NSC 95087, Py-tetrahydroserpentine, Ranitol, Raubaserp, Raubasil, Raubasin, Raumalina, Rauvasan, Sarpan, Tensyl, Tetrahydroserpentine, Vincain, Vincaine, Vincein, Vinceine, δ-Yohimbine
SMILES COC(=O)C1=CO[C@@H](C)[C@H]2CN3CCc4c([nH]c5ccccc45)[C@@H]3C[C@H]12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H24N2O3/c1-12-16-10-23-8-7-14-13-5-3-4-6-18(13)22-20(14)19(23)9-15(16)17(11-26-12)21(24)25-2/h3-6,11-12,15-16,19,22H,7-10H2,1-2H3/t12-,15-,16+,19-/m0/s1
Formula Weight 352.1787
Chemical Name or Material Ajmalicine
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.