missing translation for 'onlineSavingsMsg'
Learn More

Temsirolimus Acetonide, TRC

Product Code. 30410814
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410814 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410814 Supplier Toronto Research Chemicals Supplier No. TRCT01797050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Temsirolimus Acetonide
CAS 162635-03-2
Molecular Formula C59 H91 N O16
Quantity 50 mg
InChI Formula InChI=1S/C59H91NO16/c1-35-19-15-14-16-20-36(2)47(69-11)31-43-24-22-41(7)59(68,76-43)53(64)54(65)60-26-18-17-21-44(60)55(66)74-48(32-45(61)37(3)28-40(6)51(63)52(71-13)50(62)39(5)27-35)38(4)29-42-23-25-46(49(30-42)70-12)75-56(67)58(10)33-72-57(8,9)73-34-58/h14-16,19-20,28,35,37-39,41-44,46-49,51-52,63,68H,17-18,21-27,29-34H2,1-13H3/b16-14+,19-15+,36-20+,40-28+/t35-,37-,38-,39-,41-,42+,43+,44+,46-,47+,48+,49-,51-,52+,59-/m1/s1
SMILES CO[C@@H]1C[C@H](C[C@@H](C)[C@@H]2CC(=O)[C@H](C)\C=C(/C)\[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C\C=C(/C)\[C@H](C[C@@H]3CC[C@@H](C)[C@@](O)(O3)C(=O)C(=O)N4CCCC[C@H]4C(=O)O2)OC)CC[C@H]1OC(=O)C5(C)COC(C)(C)OC5
Molecular Weight (g/mol) 1070.35
Formula Weight 1069.6338
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.