missing translation for 'onlineSavingsMsg'
Learn More

5-(Thiophen-2-yl)isoxazole-3-carboxylic Acid, TRC

Product Code. 30411222
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411222 1 g -
30422606 500 mg -
30411894 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411222 Supplier Toronto Research Chemicals Supplier No. TRCT2667911G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 763109-71-3
Molecular Formula C8H5NO3S
Molecular Weight (g/mol) 195.19
Synonym 5-(2-Thienyl)-3-isoxazolecarboxylic Acid
IUPAC Name 5-thiophen-2-yl-1,2-oxazole-3-carboxylic acid
SMILES OC(=O)c1cc(on1)c2cccs2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H5NO3S/c10-8(11)5-4-6(12-9-5)7-2-1-3-13-7/h1-4H,(H,10,11)
Formula Weight 194.999
Chemical Name or Material 5-(Thiophen-2-yl)isoxazole-3-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.