missing translation for 'onlineSavingsMsg'
Learn More

2'-Phenoxymethanesulfonanilide, TRC

Product Code. 30411918
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411918 1 g -
30422973 25 g 25g
30411592 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411918 Supplier Toronto Research Chemicals Supplier No. TRCP3186181G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51765-51-6
Molecular Formula C13 H13 N O3 S
Molecular Weight (g/mol) 263.31
Synonym N-(2-Phenoxyphenyl)methanesulphonamide, Nimesulide Imp. B (EP)
IUPAC Name N-(2-phenoxyphenyl)methanesulfonamide
SMILES CS(=O)(=O)Nc1ccccc1Oc2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H13NO3S/c1-18(15,16)14-12-9-5-6-10-13(12)17-11-7-3-2-4-8-11/h2-10,14H,1H3
Formula Weight 263.0616
Chemical Name or Material N-(2-Phenoxyphenyl)methanesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.