missing translation for 'onlineSavingsMsg'
Learn More

Nutriacholic Acid Methyl Ester, TRC

Product Code. 30412001
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412001 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412001 Supplier Toronto Research Chemicals Supplier No. TRCN9255555G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nutriacholic Acid Methyl Ester
CAS 10538-59-7
Molecular Formula C25H40O4
Quantity 5 g
Synonym 3alpha-Hydroxy-7-oxo-5beta-cholan-24-oic Acid Methyl Ester, 3alpha-Hydroxy-7-oxo-5beta-cholanic Acid Methyl Ester, 7-Ketolithocholic Acid Methyl Ester, Methyl 3alpha-Hydroxy-7-oxo-5beta-cholan-24-oate, Methyl 3alpha-Hydroxy-7-oxo-5beta-cholanoate, Methyl 7-Keto-3alpha-hydroxy-5beta-cholanoate
InChI Formula InChI=1S/C25H40O4/c1-15(5-8-22(28)29-4)18-6-7-19-23-20(10-12-25(18,19)3)24(2)11-9-17(26)13-16(24)14-21(23)27/h15-20,23,26H,5-14H2,1-4H3/t15-,16+,17-,18-,19+,20+,23+,24+,25-/m1/s1
SMILES COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@H]3[C@H](CC[C@]12C)[C@@]4(C)CC[C@@H](O)C[C@H]4CC3=O
IUPAC Name methyl (4R)-4-[(3R,5S,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-7-oxo-1,2,3,4,5,6,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoate
Molecular Weight (g/mol) 404.58
Formula Weight 404.2927
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.