missing translation for 'onlineSavingsMsg'
Learn More

Rifabutin N-Oxide, TRC

Product Code. 30412299
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412299 2.5 mg 2.5mg
30415667 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30412299 Supplier Toronto Research Chemicals Supplier No. TRCR5050202.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645406-37-7
Molecular Formula C46 H62 N4 O12
Molecular Weight (g/mol) 863
Synonym Spiro[9,4-(epoxypentadeca[1,11,13]trienimino)-2H-furo[2',3':7,8]naphth[1,2-d]imidazole-2,4'-piperidine]-5,10,26(3H,9H)-trione, 16-(acetyloxy)-6,18,20-trihydroxy-14-methoxy-7,9,15,17,19,21,25-heptamethyl-1'-(2-methylpropyl)-, 1'-oxide, (9S,12E,14S,15R,16S,17R,18R,19R,20S,21S,22E,24Z)-, Rifabutin N-oxide
IUPAC Name [(7S,9E,11S,12R,13S,14R,15R,16R,17S,18S,19E,21Z)-2,15,17,32-tetrahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-1'-(2-methylpropyl)-1'-oxido-6,23-dioxospiro[8,33-dioxa-24,27,29-triazapentacyclo[23.6.1.14,7.05,31.026,30]tritriaconta-1(31),2,4,9,19,21,25(32),26,29-nonaene-28,4'-piperidin-1-ium]-13-yl] acetate
SMILES CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4C(=O)C(=C5NC6(CC[N+]([O-])(CC(C)C)CC6)N=C5c4c3C2=O)NC(=O)\C(=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(=O)C)[C@@H]1C)\C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C46H62N4O12/c1-22(2)21-50(58)18-16-46(17-19-50)48-34-31-32-39(54)28(8)42-33(31)43(56)45(10,62-42)60-20-15-30(59-11)25(5)41(61-29(9)51)27(7)38(53)26(6)37(52)23(3)13-12-14-24(4)44(57)47-36(40(32)55)35(34)49-46/h12-15,20,22-23,25-27,30,37-38,41,49,52-54H,16-19,21H2,1-11H3,(H,47,57)/b13-12+,20-15+,24-14-/t23-,25+,26+,27+,30-,37-,38+,41+,45-,46?,50?/m0/s1
Formula Weight 862.4364
Chemical Name or Material Rifabutin N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.