missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terbinafine Dimer Impurity Dihydrochloride Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Terbinafine Dimer Impurity Dihydrochloride Salt |
| Purity Grade Notes | HPLC |
| Molecular Formula | C36 H40 N2 . 2HCL |
| Quantity | 50 mg |
| Synonym | (2E,4E)-4-(4,4-Dimethyl-2-pentyn-1-ylidene)-N1,N5-dimethyl-N1,N5-bis(1-naphthalenylmethyl)-2-pentene-1,5-diamine Dihydrochloride Salt |
| InChI Formula | InChI=1S/C36H40N2.2ClH/c1-36(2,3)24-12-14-29(26-38(5)28-33-21-11-19-31-17-7-9-23-35(31)33)15-13-25-37(4)27-32-20-10-18-30-16-6-8-22-34(30)32;;/h6-11,13-23H,25-28H2,1-5H3;2*1H/b15-13+,29-14+;; |
| SMILES | Cl.Cl.CN(C\C=C\C(=C/C#CC(C)(C)C)\CN(C)Cc1cccc2ccccc12)Cc3cccc4ccccc34 |
| IUPAC Name | (E,4E)-4-(4,4-dimethylpent-2-ynylidene)-N,N'-dimethyl-N,N'-bis(1-naphthylmethyl)pent-2-ene-1,5-diamine;hydrochloride |
| Molecular Weight (g/mol) | 573.64 |
| Formula Weight | 572.2725 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?