missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nor Cholic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Nor Cholic acid |
| CAS | 60696-62-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C23 H38 O5 |
| Quantity | 100 mg |
| Synonym | 24-Norcholan-23-oic acid','3,7,12-trihydroxy-','(3α,5β,7α,12α)-, 23-Nor-5β-cholanic acid','3α,7α,12α-trihydroxy- (6CI), (3α,5β,7α,12α)-3,7,12-Trihydroxy-24-norcholan-23-oic acid, 23-Norcholic acid, 24-Norcholic acid, 3α,7α,12α-Trihydroxy-24-nor-5β-cholan-23-oic acid, Norcholic acid |
| InChI Formula | InChI=1S/C23H38O5/c1-12(8-20(27)28)15-4-5-16-21-17(11-19(26)23(15,16)3)22(2)7-6-14(24)9-13(22)10-18(21)25/h12-19,21,24-26H,4-11H2,1-3H3,(H,27,28)/t12-,13+,14-,15-,16+,17+,18-,19+,21+,22+,23-/m1/s1 |
| SMILES | C[C@H](CC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C |
| IUPAC Name | (3R)-3-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]butanoic acid |
| Molecular Weight (g/mol) | 394.54 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?