missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4’-Oxydibenzenesulfonyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 4,4'-Oxydibenzenesulfonyl Chloride |
| CAS | 121-63-1 |
| Molecular Formula | C12H8Cl2O5S2 |
| Quantity | 25 g |
| Synonym | 4,4'-Oxybis[benzenesulfonyl Chloride], 4,4'-Bis(chlorosulfonyl)biphenyl Ether, 4,4'-Bis(chlorosulfonyl)diphenyl Ether, 4,4'-Oxybis(benzenesulfonyl Chloride), 4,4'-Oxydibenzenesulfonyl Chloride, Bis(4-Chlorosulfonylphenyl) Ether, Diphenyl Ether 4,4’-Disulfonyl Chloride, NSC 212, Oxybis(4-benzenesulfonyl Chloride), Oxybis(benzenesulfonyl Chloride), Phenoxybenzene-4,4'-disulfonyl Chloride, p,p'-Oxybis(benzenesulfonyl Chloride), 4,4’-Oxybisbenzenesulfonyl Chloride |
| InChI Formula | InChI=1S/C12H8Cl2O5S2/c13-20(15,16)11-5-1-9(2-6-11)19-10-3-7-12(8-4-10)21(14,17)18/h1-8H |
| SMILES | ClS(=O)(=O)c1ccc(Oc2ccc(cc2)S(=O)(=O)Cl)cc1 |
| IUPAC Name | 4-(4-chlorosulfonylphenoxy)benzenesulfonyl chloride |
| Molecular Weight (g/mol) | 367.22 |
| Formula Weight | 365.919 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?