missing translation for 'onlineSavingsMsg'
Learn More

Pipazetate, TRC

Product Code. 30413768
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413768 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413768 Supplier Toronto Research Chemicals Supplier No. TRCP475705100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pipazetate
CAS 2167-85-3
Molecular Formula C21 H25 N3 O3 S
Quantity 100 mg
Synonym 10H-Pyrido[3,2-b][1,4]benzothiazine-10-carboxylic acid','2-[2-(1-piperidinyl)ethoxy]ethyl ester, 10H-Pyrido[3,2-b][1,4]benzothiazine-10-carboxylic acid','2-(2-piperidinoethoxy)ethyl ester (7CI,8CI), Ethanol','2-(2-piperidinoethoxy)-','10H-pyrido[3,2-b][1,4]benzothiazine-10-carboxylate (ester) (8CI), 2-(2-Piperidinoethoxy)ethyl 1-azaphenothiazine-10-carboxylate, 2-(2-Piperidinoethoxy)ethyl 10H-pyrido[3,2-b][1,4]benzothiazine-10-carboxylate, D 254, Pipazetate, Pipazethate, SKF 70230A, SQ 15874
InChI Formula InChI=1S/C21H25N3O3S/c25-21(27-16-15-26-14-13-23-11-4-1-5-12-23)24-17-7-2-3-8-18(17)28-19-9-6-10-22-20(19)24/h2-3,6-10H,1,4-5,11-16H2
SMILES O=C(OCCOCCN1CCCCC1)N2c3ccccc3Sc4cccnc24
IUPAC Name 2-(2-piperidin-1-ylethoxy)ethyl pyrido[3,2-b][1,4]benzothiazine-10-carboxylate
Molecular Weight (g/mol) 399.51
Formula Weight 399.1617
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.