missing translation for 'onlineSavingsMsg'
Learn More

Spinosyn B, TRC

Product Code. 30413835
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413835 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30413835 Supplier Toronto Research Chemicals Supplier No. TRCS6819605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Spinosyn B
CAS 131929-61-8
Molecular Formula C40 H63 N O10
Quantity 5 mg
Synonym A 83543B
InChI Formula InChI=1S/C40H63NO10/c1-9-25-11-10-12-33(51-35-16-15-32(41-5)22(3)47-35)21(2)36(43)31-19-29-27(30(31)20-34(42)49-25)14-13-24-17-26(18-28(24)29)50-40-39(46-8)38(45-7)37(44-6)23(4)48-40/h13-14,19,21-30,32-33,35,37-41H,9-12,15-18,20H2,1-8H3/t21-,22-,23+,24-,25+,26-,27-,28-,29-,30+,32+,33+,35+,37+,38-,39-,40+/m1/s1
SMILES CC[C@H]1CCC[C@H](O[C@H]2CC[C@H](NC)[C@@H](C)O2)[C@@H](C)C(=O)C3=C[C@H]4[C@@H]5C[C@@H](C[C@H]5C=C[C@H]4[C@@H]3CC(=O)O1)O[C@@H]6O[C@@H](C)[C@H](OC)[C@@H](OC)[C@H]6OC
Molecular Weight (g/mol) 717.929
Formula Weight 717.445
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.