missing translation for 'onlineSavingsMsg'
Learn More

Pentaethylene glycol dichloride, TRC

Product Code. 30414670
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414670 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414670 Supplier Toronto Research Chemicals Supplier No. TRCP270230250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pentaethylene glycol dichloride
CAS 5197-65-9
Molecular Formula C10H20Cl2O4
Quantity 250 mg
Synonym 1,14-Dichloro-3,6,9,12-tetraoxatetradecane, 1,2-bis[2-(2-Chloroethoxy)ethoxy]-ethane, 1,14-Dichloro-3,6,9,12-tetraoxatetradecane
InChI Formula InChI=1S/C10H20Cl2O4/c11-1-3-13-5-7-15-9-10-16-8-6-14-4-2-12/h1-10H2
SMILES ClCCOCCOCCOCCOCCCl
IUPAC Name 1,2-bis[2-(2-chloroethoxy)ethoxy]ethane
Molecular Weight (g/mol) 275.17
Formula Weight 274.0739
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.