missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-Alpha-(1-naphthalenylmethyl)-2-furanpropanoic Acid, TRC

Product Code. 30414687
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30414687 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414687 Supplier Toronto Research Chemicals Supplier No. TRCT293510250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25379-26-4
Molecular Formula C18 H20 O3
Quantity 250 mg
Synonym Tetrahydro-alpha-(1-naphthalenylmethyl)-2-furanpropanoic acid, 2-Furanpropionic acid, tetrahydro-alpha-(1-naphthylmethyl)- (7CI,8CI), LS 74, Tetrahydro-alpha-(1-naphthylmethyl)-2-furanpropionic acid, alpha-(1-Naphthalenylmethyl)-3-tetrahydrofuran-2-propionic acid
InChI Formula InChI=1S/C18H20O3/c19-18(20)15(12-16-8-4-10-21-16)11-14-7-3-6-13-5-1-2-9-17(13)14/h1-3,5-7,9,15-16H,4,8,10-12H2,(H,19,20)
SMILES OC(=O)C(CC1CCCO1)Cc2cccc3ccccc23
IUPAC Name 2-(naphthalen-1-ylmethyl)-3-(oxolan-2-yl)propanoic acid
Molecular Weight (g/mol) 284.35
Formula Weight 284.1412
Chemical Name or Material 2-[(Naphthalen-1-yl)methyl]-3-(tetrahydrofuran-2-yl)propanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.