missing translation for 'onlineSavingsMsg'
Learn More

Telmisartan Amide, TRC

Product Code. 30414695
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30414695 1 g
30424706 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30414695 Supplier Toronto Research Chemicals Supplier No. TRCT0170201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 915124-86-6
Molecular Formula C33 H31 N5 O
Molecular Weight (g/mol) 513.63
Synonym 4'-[[4-Methyl-6-(1-methyl-1H-benz-imidazol-2-yl)-2-propyl-1H-benzimidazol-1-yl]-methyl]biphenyl-2-carboxamide, 4'-[(1,4'-Dimethyl-2'-propyl[2,6'-bi-1H-benzimidazol]-1'-yl)methyl][1,1'-biphenyl]-2-carboxamide, [1,1'-Biphenyl]-2-carboxamide, 4'-[(1,4'-dimethyl-2'-propyl[2,6'-bi-1H-benzimidazol]-1'-yl)methyl]-, Telmisartan Imp F (EP)
IUPAC Name 2-[4-[[4-methyl-6-(1-methylbenzimidazol-2-yl)-2-propylbenzimidazol-1-yl]methyl]phenyl]benzamide
SMILES CCCc1nc2c(C)cc(cc2n1Cc3ccc(cc3)c4ccccc4C(=O)N)c5nc6ccccc6n5C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C33H31N5O/c1-4-9-30-36-31-21(2)18-24(33-35-27-12-7-8-13-28(27)37(33)3)19-29(31)38(30)20-22-14-16-23(17-15-22)25-10-5-6-11-26(25)32(34)39/h5-8,10-19H,4,9,20H2,1-3H3,(H2,34,39)
Formula Weight 513.2529
Percent Purity >95%
Chemical Name or Material Telmisartan Amide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.