missing translation for 'onlineSavingsMsg'
Learn More

Phenelzine Sulfate, TRC

Product Code. 30414891
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414891 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414891 Supplier Toronto Research Chemicals Supplier No. TRCP2959005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenelzine Sulfate
CAS 156-51-4
Purity Grade Notes HPLC
Molecular Formula C8 H12 N2 . H2 O4 S
Quantity 5 g
Synonym Hydrazine','(2-phenylethyl)-','sulfate (1:1), Hydrazine','phenethyl-','sulfate (1:1) (8CI), (2-Phenylethyl)hydrazine dihydrogen sulfate, (β-Phenylethyl)hydrazine dihydrogen sulfate, (β-Phenylethyl)hydrazine sulfate, 2-Phenethylhydrazine sulfate, Estinerval, Fenelzin, Kalgan, Mao-rem, NSC 170957, Nardelzine, Nardil, Phenalzine dihydrogen sulfate, Phenelzine acid sulfate, Phenelzine bisulfate, Phenelzine dihydrogen sulfate, Phenelzine hydrogen sulfate, Phenelzine sulfate, Phenethylhydrazine sulfate, Stinerval, W 1544A
InChI Formula InChI=1S/C8H12N2.H2O4S/c9-10-7-6-8-4-2-1-3-5-8;1-5(2,3)4/h1-5,10H,6-7,9H2;(H2,1,2,3,4)
SMILES NNCCc1ccccc1.OS(=O)(=O)O
IUPAC Name 2-phenylethylhydrazine;sulfuric acid
Molecular Weight (g/mol) 234.27
Formula Weight 234.0674
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.