missing translation for 'onlineSavingsMsg'
Learn More

3-Oxo-12alpha-hydroxy-5beta-cholanoic Acid, TRC

Product Code. 30416396
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416396 2.5 g 2.5g
30424206 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30416396 Supplier Toronto Research Chemicals Supplier No. TRCO8568702.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4185-01-7
Molecular Formula C24 H38 O4
Molecular Weight (g/mol) 390.56
Synonym Cholan-24-oic acid, 12-hydroxy-3-oxo-, (5β,12α)-, 5β-Cholan-24-oic acid, 12α-hydroxy-3-oxo- (8CI), 5β-Cholanic acid, 12α-hydroxy-3-oxo- (7CI), (5β,12α)-12-Hydroxy-3-oxocholan-24-oic acid, 12α-Hydroxy-3-keto-5β-cholanoic acid, 12α-Hydroxy-3-oxo-5β-chol-24-oic acid, 12α-Hydroxy-3-oxo-5β-cholan-24-oic acid, 12α-Hydroxy-3-oxo-5β-cholanic acid, 12α-Hydroxy-3-oxo-5β-cholanoic acid, 3-Oxo-12α-hydroxy-5β-cholanic acid, 3-Oxo-12α-hydroxy-5β-cholanoic acid, 3-Oxo-12α-hydroxycholan-24-oic acid, 3-Oxodeoxycholic acid
IUPAC Name (4R)-4-[(5R,8R,9S,10S,12S,13R,14S,17R)-12-hydroxy-10,13-dimethyl-3-oxo-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4CC(=O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C24H38O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-15,17-21,26H,4-13H2,1-3H3,(H,27,28)/t14-,15-,17+,18-,19+,20+,21+,23+,24-/m1/s1
Formula Weight 390.277
Percent Purity >95%
Chemical Name or Material 3-Oxo-12alpha-hydroxy-5beta-cholanoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.