missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate-13C,D3, TRC

Product Code. 30418444
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418444 10 mg 10mg
30410474 50 mg 50mg
30423321 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30418444 Supplier Toronto Research Chemicals Supplier No. TRCS96021010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102212-93-1
Molecular Formula C13CD3NaO2
Molecular Weight (g/mol) 86.04
Synonym Acetic-1-13C-2,2,2-d3 Acid Sodium Salt
IUPAC Name sodium;2,2,2-trideuterioacetate
SMILES [2H]C([2H])([13C](O[Na])=O)[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1/i1D3,2+1;
Formula Weight 86.0253
Chemical Name or Material Sodium Acetate-13C,D3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.