missing translation for 'onlineSavingsMsg'
Learn More

Pyridoxine Dimer (80%), TRC

Product Code. 30419299
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
50 mg
Unit Size:
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419299 2.5 mg 2.50mg
30423103 50 mg -
30425615 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30419299 Supplier Toronto Research Chemicals Supplier No. TRCP9912252.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19203-56-6
Molecular Formula C16H20N2O5
Molecular Weight (g/mol) 320.34
Synonym 5-Hydroxy-2-[[3-hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridyl]methyl]-6-methyl-3,4-pyridinedimethanol
IUPAC Name 6-[[3-hydroxy-5-(hydroxymethyl)-2-methylpyridin-4-yl]methyl]-4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol
SMILES OCC1=C(CC2=NC(C)=C(O)C(CO)=C2CO)C(O)=C(C)N=C1

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C16H20N2O5/c1-8-15(22)11(10(5-19)4-17-8)3-14-12(6-20)13(7-21)16(23)9(2)18-14/h4,19-23H,3,5-7H2,1-2H3
Formula Weight 320.1372
Chemical Name or Material 6-[(3-hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridyl)methyl]-4,5-bis(hydroxymethyl)-2-methyl-pyridin-3-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.