missing translation for 'onlineSavingsMsg'
Learn More

Rapidosept-d2, TRC

Product Code. 30419482
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30419482 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30419482 Supplier Toronto Research Chemicals Supplier No. TRCR124502100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Rapidosept-d2
CAS 883001-15-8
Molecular Formula C7H4D2Cl2O
Quantity 100 mg
Synonym 2,4-Dichloro-benzenemethanol-d2, 2,4-Dichloro-benzyl Alcohol-d2, 2,4-Dichlorobenzenemethanol-d2, 2,4-Dichlorobenzyl Alcohol-d2, Dybenal-d2, Myacide-d2, Myacide SP-d2, NSC 15635-d2
InChI Formula InChI=1S/C7H6Cl2O/c8-6-2-1-5(4-10)7(9)3-6/h1-3,10H,4H2/i4D2
SMILES [2H]C([2H])(O)c1ccc(Cl)cc1Cl
IUPAC Name dideuterio-(2,4-dichlorophenyl)methanol
Molecular Weight (g/mol) 179.04
Formula Weight 177.9921
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.