missing translation for 'onlineSavingsMsg'
Learn More

Saclofen, TRC

Product Code. 30422178
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422178 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422178 Supplier Toronto Research Chemicals Supplier No. TRCS0808505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Saclofen
CAS 125464-42-8
Molecular Formula C9 H12 Cl N O3 S
Quantity 5 mg
Synonym Saclofen, β-(Aminomethyl)-4-chlorobenzeneethanesulfonic Acid
InChI Formula InChI=1S/C9H12ClNO3S/c10-9-3-1-7(2-4-9)8(5-11)6-15(12,13)14/h1-4,8H,5-6,11H2,(H,12,13,14)
SMILES NCC(CS(=O)(=O)O)c1ccc(Cl)cc1
IUPAC Name 3-amino-2-(4-chlorophenyl)propane-1-sulfonic acid
Molecular Weight (g/mol) 249.714
Formula Weight 249.023
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.