missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol Dicaprate, TRC

Product Code. 30422699
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422699 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422699 Supplier Toronto Research Chemicals Supplier No. TRCP79714550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propylene Glycol Dicaprate
CAS 53824-77-4
Molecular Formula C23 H44 O4
Quantity 50 mg
Synonym Propylene Glycol Dicaprate, 1,2-Propanediol Didecanoate, Captex 100, Nikkol PDD, Propylene glycol didecanoate
InChI Formula InChI=1S/C23H44O4/c1-4-6-8-10-12-14-16-18-22(24)26-20-21(3)27-23(25)19-17-15-13-11-9-7-5-2/h21H,4-20H2,1-3H3
SMILES CCCCCCCCCC(=O)OCC(C)OC(=O)CCCCCCCCC
IUPAC Name 2-decanoyloxypropyl decanoate
Molecular Weight (g/mol) 384.593
Formula Weight 384.324
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.