missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sparfloxacin, TRC
Specifications
Specifications
| Chemical Name or Material | Sparfloxacin |
| CAS | 110871-86-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H22 F2 N4 O3 |
| Quantity | 100 mg |
| Synonym | Sparfloxacin, 5-Amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethylpiperazin-1-yl]-6,8-difluoro-4-oxo-quinoline-3-carboxylic acid, 3-Quinolinecarboxylic acid','5-amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethyl-1-piperazinyl]-6,8-difluoro-1,4-dihydro-4-oxo-','rel-, 3-Quinolinecarboxylic acid','5-amino-1-cyclopropyl-7-(3,5-dimethyl-1-piperazinyl)-6,8-difluoro-1,4-dihydro-4-oxo-','cis-, rel-5-Amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethyl-1-piperazinyl]-6,8-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid, AT 4140, CI 978, CP 103826, PD 1315-1, PD 131501, Parox, Salocin 120, Spara, Sparcin, Zagam |
| InChI Formula | InChI=1S/C19H22F2N4O3/c1-8-5-24(6-9(2)23-8)17-13(20)15(22)12-16(14(17)21)25(10-3-4-10)7-11(18(12)26)19(27)28/h7-10,23H,3-6,22H2,1-2H3,(H,27,28)/t8-,9+ |
| SMILES | C[C@@H]1CN(C[C@H](C)N1)c2c(F)c(N)c3C(=O)C(=CN(C4CC4)c3c2F)C(=O)O |
| IUPAC Name | 5-amino-1-cyclopropyl-7-[(3R,5S)-3,5-dimethylpiperazin-1-yl]-6,8-difluoro-4-oxoquinoline-3-carboxylic acid |
| Molecular Weight (g/mol) | 392.4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?