missing translation for 'onlineSavingsMsg'
Learn More

Sangivamycin, TRC

Product Code. 30423286
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423286 10 mg 10mg
30426862 1 mg -
30418022 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423286 Supplier Toronto Research Chemicals Supplier No. TRCS11250310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18417-89-5
Molecular Formula C12H15N5O5
Molecular Weight (g/mol) 309.28
Synonym 4-Amino-7-beta-D-ribofuranosyl-7H-pyrrolo[2,3-d]pyrimidine-5-carboxamide, 7-Deazaadenosine-7-carboxamide, NSC 65346
IUPAC Name 3,4,7,8-tetramethyl-1,10-phenanthroline
SMILES Cc1cnc2c(ccc3c(C)c(C)cnc23)c1C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H16N2/c1-9-7-17-15-13(11(9)3)5-6-14-12(4)10(2)8-18-16(14)15/h5-8H,1-4H3
Formula Weight 236.1313
Chemical Name or Material Sangivamycin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.