missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-Hydroxy-androst-5-ene-17-methylsulfate, TRC

Product Code. 30424971
Change view
Click to view available options
Quantity:
0.5 mg
2.5 mg
5 mg
Unit Size:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424971 5 mg -
30412054 0.5 mg 0.5mg
30420973 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424971 Supplier Toronto Research Chemicals Supplier No. TRCS3333005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131211-37-5
Molecular Formula C20H31NaO5S
Molecular Weight (g/mol) 406.51
Synonym Androst-5-ene-17-methanol, 3-hydroxy-, α-(hydrogen sulfate), monosodium salt, (3β,17β)- (9CI), (3β,17β)-3-Hydroxyandrost-5-ene-17-methanol α-(hydrogen sulfate) monosodium salt, Sodium 3β-Hydroxyandrost-5-ene-17β-methyl sulfate, 3β-Hydroxyandrost-5-ene-17β-methyl sodium sulfate
SMILES O[C@H]1CC[C@]2(C)[C@@]3([H])CC[C@]4(C)[C@@H](COS(=O)(O[Na])=O)CC[C@@]4([H])[C@]3([H])CC=C2C1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H32O5S.Na/c1-19-9-7-15(21)11-13(19)3-5-16-17-6-4-14(12-25-26(22,23)24)20(17,2)10-8-18(16)19;/h3,14-18,21H,4-12H2,1-2H3,(H,22,23,24);/q;+1/p-1/t14-,15+,16+,17+,18+,19+,20-;/m1./s1
Formula Weight 406.179
Chemical Name or Material Sodium 3-Hydroxy-androst-5-ene-17-methylsulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.