missing translation for 'onlineSavingsMsg'
Learn More

N6-{[(2-Phenylethyl)amino]carbonothioyl}lysine (PEITC-Lys), TRC

Product Code. 30425162
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425162 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425162 Supplier Toronto Research Chemicals Supplier No. TRCP335918100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N6-{[(2-Phenylethyl)amino]carbonothioyl}lysine (PEITC-Lys)
CAS 1211456-36-8
Molecular Formula C15 H23 N3 O2 S
Quantity 100 mg
Synonym N6-[[(2-Phenylethyl)amino]thioxomethyl]-L-lysine
InChI Formula InChI=1S/C15H23N3O2S/c16-13(14(19)20)8-4-5-10-17-15(21)18-11-9-12-6-2-1-3-7-12/h1-3,6-7,13H,4-5,8-11,16H2,(H,19,20)(H2,17,18,21)/t13-/m0/s1
SMILES OC([C@H](CCCCNC(NCCC1=CC=CC=C1)=S)N)=O
IUPAC Name N6-(phenethylcarbamothioyl)-L-lysine
Molecular Weight (g/mol) 309.43
Formula Weight 309.15
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.