missing translation for 'onlineSavingsMsg'
Learn More

Pentadecafluorooctanoic Acid Ammonium Salt, TRC

Product Code. 30427296
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427296 2.5 g 2.5g
30428405 5 g 5g
30417016 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427296 Supplier Toronto Research Chemicals Supplier No. TRCP2681002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3825-26-1
Molecular Formula C8 H F15 O2 . H3 N
Molecular Weight (g/mol) 431.1
Synonym Perfluorooctanoic Acid Ammonium, Ammonium perfluorooctanoate, Octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, ammonium salt (1:1), Octanoic acid, pentadecafluoro-, ammonium salt (8CI, 9CI), Ammonium pentadecafluorooctanoate, Ammonium perfluorocaprylate, Ammonium perfluorooctanoate, Ammonium perfluorooctoate, CXR 1002, DS 101, FC 1015, FC 143, Fluorad FC 143, FX 1006, Pentadecafluorooctanoic acid ammonium salt, Perfluorooctanoic acid ammonium salt, Unidyne DS 101
IUPAC Name azanium;2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
SMILES [NH4+].[O-]C(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C8HF15O2.H3N/c9-2(10,1(24)25)3(11,12)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)23;/h(H,24,25);1H3
Formula Weight 431.0003
Percent Purity >95%
Chemical Name or Material Perfluorooctanoic acid ammonium salt PFOA POAA
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.