missing translation for 'onlineSavingsMsg'
Learn More

Taurolidine-D6, TRC

Product Code. 30427526
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427526 1 mg
30411128 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30427526 Supplier Toronto Research Chemicals Supplier No. TRCT0090521MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C7 D6 H10 N4 O4 S2
Molecular Weight (g/mol) 290.393
Synonym 4,4'-Methylenebis[tetrahydro-2H-1,2,4-thiadiazine 1,1,1',1'-Tetraoxide-D6, ¡4,4'-Methylenebis(perhydro-1,2,4-thiadiazin 1,1-dioxide)-D6, Taurolin-D6, Tauroline-D6
IUPAC Name 3,3-dideuterio-4-[dideuterio-(3,3-dideuterio-1,1-dioxo-1,2,4-thiadiazinan-4-yl)methyl]-1,2,4-thiadiazinane 1,1-dioxide
SMILES [2H]C([2H])(N1CCS(=O)(=O)NC1([2H])[2H])N2CCS(=O)(=O)NC2([2H])[2H]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C7H16N4O4S2/c12-16(13)3-1-10(5-8-16)7-11-2-4-17(14,15)9-6-11/h8-9H,1-7H2/i5D2,6D2,7D2
Formula Weight 290.099
Chemical Name or Material Taurolidine-D6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.