missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate-d3, TRC

Product Code. 30428572
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428572 5 g 5g
30424477 2.5 g 2.5g
30418890 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30428572 Supplier Toronto Research Chemicals Supplier No. TRCS6109735G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39230-37-0
Molecular Formula C2 D3 H O2 . C2 D3 O2 . Na
Molecular Weight (g/mol) 148.123
Synonym Acetic-d3 acid, sodium salt (2:1) (9CI), Sodium hydrogen di(acetate-d3), Sodium hydrogen di(acetate-D3), Acetic acid-d3 hemisodium salt
IUPAC Name sodium;2,2,2-trideuterioacetate;2,2,2-trideuterioacetic acid
SMILES [Na+].[2H]C([2H])([2H])C(=O)O.[2H]C([2H])([2H])C(=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/2C2H4O2.Na/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+1/p-1/i2*1D3;
Formula Weight 148.062
Chemical Name or Material Sodium Acetate-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.