missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quizalofop-P, TRC
Specifications
Specifications
| Chemical Name or Material | Quizalofop-P (free acid) |
| CAS | 94051-08-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 H13 Cl N2 O4 |
| Quantity | 25 mg |
| Synonym | Propanoic acid','2-[4-[(6-chloro-2-quinoxalinyl)oxy]phenoxy]-','(2R)-, Propanoic acid','2-[4-[(6-chloro-2-quinoxalinyl)oxy]phenoxy]-','(R)-, (2R)-2-[4-[(6-Chloro-2-quinoxalinyl)oxy]phenoxy]propanoic acid, (R)-(+)-Quizalofop, DPX-Y 6202-31, Propaquizafop free acid, Quizalofop-P, R-Quizalofop, R-Quizalofop-acid, Quizalofop-P (free acid), (R)-Quizalofop |
| InChI Formula | InChI=1S/C17H13ClN2O4/c1-10(17(21)22)23-12-3-5-13(6-4-12)24-16-9-19-15-8-11(18)2-7-14(15)20-16/h2-10H,1H3,(H,21,22)/t10-/m1/s1 |
| SMILES | C[C@@H](Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1)C(=O)O |
| IUPAC Name | (2R)-2-[4-(6-chloroquinoxalin-2-yl)oxyphenoxy]propanoic acid |
| Molecular Weight (g/mol) | 344.75 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?