missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uridine 5’-Diphospho-N-acetylglucosamine Disodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Uridine 5'-Diphospho-N-acetylglucosamine Disodium Salt |
| CAS | 91183-98-1 |
| Molecular Formula | C17 H25 N3 O17 P2 . 2 Na |
| Quantity | 50 mg |
| Synonym | Uridine 5'-(trihydrogen diphosphate)','P'-[2-(acetylamino)-2-deoxy-α-D-glucopyranosyl] ester','sodium salt (1:2), Uridine 5'-(trihydrogen diphosphate)','P'-[2-(acetylamino)-2-deoxy-α-D-glucopyranosyl] ester','disodium salt (9CI), Uridine 5'-(trihydrogen diphosphate) P'-[2-(acetylamino)-2-deoxy-α-D-glucopyranosyl] ester disodium salt, Uridine 5'-diphospho-N-acetylglucosamine disodium salt, UDP-N-acetyl-D-glucosamine disodium salt, UDP-N-acetylglucosamine disodium salt, UDP-GlcNAc disodium salt, UDP-α-D-N-acetylglucosamine disodium salt, Uridine 5'-(2-acetamido-2-deoxy-α-D-glucosyl pyrophosphate) disodium salt |
| InChI Formula | InChI=1S/C17H27N3O17P2.2Na/c1-6(22)18-10-13(26)11(24)7(4-21)35-16(10)36-39(31,32)37-38(29,30)33-5-8-12(25)14(27)15(34-8)20-3-2-9(23)19-17(20)28;;/h2-3,7-8,10-16,21,24-27H,4-5H2,1H3,(H,18,22)(H,29,30)(H,31,32)(H,19,23,28);;/q;2*+1/p-2/t7-,8-,10-,11-,12-,13-,14-,15-,16-;;/m1../s1 |
| SMILES | [Na+].[Na+].CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@H]1OP(=O)([O-])OP(=O)([O-])OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N3C=CC(=O)NC3=O |
| IUPAC Name | disodium;[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl] [[(2R,3S,4R,5R)-5-(2,4-dioxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] phosphate |
| Molecular Weight (g/mol) | 651.32 |
| Formula Weight | 651.0455 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?