missing translation for 'onlineSavingsMsg'
Learn More

T6167923, TRC

Product Code. 30450980
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450980 50 mg 50mg
30455964 10 mg 10mg
30450895 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450980 Supplier Toronto Research Chemicals Supplier No. TRCT96633850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1090528-71-4
Molecular Formula C17H20BrN3O3S2
Molecular Weight (g/mol) 458.39
Synonym 4-((3-Bromophenyl)sulfonyl)-N-(1-(thiophen-2-yl)ethyl)piperazine-1-carboxamide
IUPAC Name 4-(3-bromophenyl)sulfonyl-N-(1-thiophen-2-ylethyl)piperazine-1-carboxamide
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[O-]P(OC)(OC)=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H20BrN3O3S2/c1-13(16-6-3-11-25-16)19-17(22)20-7-9-21(10-8-20)26(23,24)15-5-2-4-14(18)12-15/h2-6,11-13H,7-10H2,1H3,(H,19,22)
Formula Weight 457.0129
Chemical Name or Material T6167923
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.