missing translation for 'onlineSavingsMsg'
Learn More

Ziprasidone Sulfoxide, TRC

Product Code. 30451959
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30451959 50 mg 50mg
30457237 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30451959 Supplier Toronto Research Chemicals Supplier No. TRCZ48501550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188797-80-0
Molecular Formula C21H21ClN4O2S
Molecular Weight (g/mol) 428.94
Synonym 2H-Indol-2-one, 6-chloro-1,3-dihydro-5-[2-[4-(1-oxido-1,2-benzisothiazol-3-yl)-1-piperazinyl]ethyl]-, 6-Chloro-1,3-dihydro-5-[2-[4-(1-oxido-1,2-benzisothiazol-3-yl)-1-piperazinyl]ethyl]-2H-indol-2-one, Ziprasidone sulfoxide
SMILES Clc1cc2NC(=O)Cc2cc1CCN3CCN(CC3)C4=NS(=O)c5ccccc45

Specifications

Quantity 50 mg
Formula Weight 428.1074
Chemical Name or Material Ziprasidone Sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.