missing translation for 'onlineSavingsMsg'
Learn More

Tetrathiafulvalene - 7,7,8,8-Tetracyanoquinodimethane Complex, TRC

Product Code. 30453916
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453916 10 mg 10mg
30456434 100 mg 100mg
30450673 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453916 Supplier Toronto Research Chemicals Supplier No. TRCT31096510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40210-84-2
Molecular Formula C6H4S4 . C12H4N4
Molecular Weight (g/mol) 408.54
IUPAC Name 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile;2-(1,3-dithiol-2-ylidene)-1,3-dithiole
SMILES N#CC(=C1C=CC(=C(C#N)C#N)C=C1)C#N.S2C=CSC2=C3SC=CS3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H4N4.C6H4S4/c13-5-11(6-14)9-1-2-10(4-3-9)12(7-15)8-16;1-2-8-5(7-1)6-9-3-4-10-6/h1-4H;1-4H
Formula Weight 407.9632
Chemical Name or Material Tetrathiafulvalene - 7,7,8,8-Tetracyanoquinodimethane Complex
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.