missing translation for 'onlineSavingsMsg'
Learn More

(1R,3R)-rel-3-[(1Z)-2-Chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethylcyclopropanecarboxylic Acid (2,3,4,5-Tetrafluoro-6-methylphenyl)methyl Ester-d5, TRC

Product Code. 30456652
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456652 5 mg 5mg
30454337 50 mg 50mg
30449089 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30456652 Supplier Toronto Research Chemicals Supplier No. TRCT4146055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1R,3R)-rel-3-[(1Z)-2-Chloro-3,3,3-trifluoro-1-propenyl]-2,2-dimethylcyclopropanecarboxylic Acid (2,3,4,5-Tetrafluoro-6-methylphenyl)methyl Ester-d5
Molecular Formula C17 D5 H9 Cl F7 O2
Quantity 5 mg
InChI Formula InChI=1S/C17H14ClF7O2/c1-6-7(12(20)14(22)13(21)11(6)19)5-27-15(26)10-8(16(10,2)3)4-9(18)17(23,24)25/h4,8,10H,5H2,1-3H3/b9-4-/t8-,10-/m0/s1/i1D3,5D2
SMILES [2H]C([2H])([2H])c1c(F)c(F)c(F)c(F)c1C([2H])([2H])OC(=O)[C@@H]2[C@H](\C=C(/Cl)\C(F)(F)F)C2(C)C
IUPAC Name [dideuterio-[2,3,4,5-tetrafluoro-6-(trideuteriomethyl)phenyl]methyl] (1R,3R)-3-[(Z)-2-chloro-3,3,3-trifluoroprop-1-enyl]-2,2-dimethylcyclopropane-1-carboxylate
Molecular Weight (g/mol) 423.764
Formula Weight 423.088
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.