missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-N-propyl Urea, Contains 40% Water, ~2% Acetic Acid, TRC

Product Code. 30464380
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464380 100 g -
30482362 1 g 1g
30470845 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30464380 Supplier Toronto Research Chemicals Supplier No. TRCN545750100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 816-57-9
Molecular Formula C4 H9 N3 O2
Molecular Weight (g/mol) 131.13
Synonym Urea, N-nitroso-N-propyl- (9CI, ACI), N-Nitroso-N-propylurea (ACI), Urea, 1-nitroso-1-propyl- (7CI, 8CI), 1-n-Propyl-1-nitrosourea, 1-Nitroso-1-propylurea, 1-Propyl-1-nitrosourea, N-(n-Propyl)-N-nitrosourea, N-Propyl-N-nitrosourea, NPU, NSC 46792
IUPAC Name 1-nitroso-1-propylurea
SMILES CCCN(N=O)C(=O)N

Specifications

Quantity 100 g
InChI Formula InChI=1S/C4H9N3O2/c1-2-3-7(6-9)4(5)8/h2-3H2,1H3,(H2,5,8)
Formula Weight 131.0695
Chemical Name or Material N-Nitroso-N-propylurea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.