missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid-13C6,d4 N-Hydroxysuccinimide Ester, TRC

Product Code. 30466726
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466726 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30466726 Supplier Toronto Research Chemicals Supplier No. TRCN4292570.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicotinic Acid-13C6','d4 N-Hydroxysuccinimide Ester
CAS 1209457-55-5
Molecular Formula 13C6 C4 D4 H4 N2 O4
Quantity 0.5 mg
Synonym Nicotinic Acid 13C6,D4 N-Hydroxysuccinimide Ester
InChI Formula InChI=1S/C10H8N2O4/c13-8-3-4-9(14)12(8)16-10(15)7-2-1-5-11-6-7/h1-2,5-6H,3-4H2/i1+1D,2+1D,5+1D,6+1D,7+1,10+1
SMILES [2H][13c]1n[13c]([2H])[13c]([13C](=O)ON2C(=O)CCC2=O)[13c]([2H])[13c]1[2H]
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2,4,5,6-tetradeuterio(2,3,4,5,6-13C5)pyridine-3-(13C)carboxylate
Molecular Weight (g/mol) 230.162
Formula Weight 230.094
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.