missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mugineic Acid Ammonium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Mugineic Acid Ammonium Salt |
| Molecular Formula | C12H20N2O8 . x(NH 3) |
| Synonym | (αS,βS,2S)-2-Carboxy-α-[[(3S)-3-carboxy-3-hydroxypropyl]amino]-β-hydroxy-1-azetidinebutanoic Acid Ammonium Salt, [2S-[1[αR*(R*),βR*],2R*]]-2-Carboxy-α-[(3-carboxy-3-hydroxypropyl)amino]-β-hydroxy-1-azetidinebutanoic Acid Ammonium Salt |
| InChI Formula | InChI=1S/C12H20N2O8.H3N/c15-7(11(19)20)1-3-13-9(12(21)22)8(16)5-14-4-2-6(14)10(17)18;/h6-9,13,15-16H,1-5H2,(H,17,18)(H,19,20)(H,21,22);1H3/t6?,7?,8-,9?;/m0./s1 |
| SMILES | O[C@H](C(O)=O)CCN[C@H](C(O)=O)[C@@H](O)CN1[C@H](C(O)=O)CC1.N |
| IUPAC Name | (S)-1-((2S,3S)-3-carboxy-3-(((S)-3-carboxy-3-hydroxypropyl)amino)-2-hydroxypropyl)azetidine-2-carboxylic acid, ammonia salt |
| Molecular Weight (g/mol) | 337.33 |
| Quantity | 2.5 mg |
| Shipping Condition | Room Temperature |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?